Learn More
Sigma Aldrich Fine Chemicals Biosciences JulaboR CORIOTM CD heating1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7429481EA
Encompass
In addition to the advantages of the CORIO C Circulator the CORIO CD Immersion Circulator can be equipped with an optional pump set for temperature control of external applications.Your advantagesPrecise temperature controlIintegrated timer function. When the set time has elapsed a signal sounds and the device switches off. Setting range 0 ... 999 minutesFor internal and external applications (accessories required)For bath tanks with a max. filling volume of 50 litersBright white easy to read displayVery quietEasy operationEasy pump change-over between internal and external circulationUSB connectionClass III (FL) according to DIN 12876-1LED Temperature Display2kW heating capacity 15 L/min Circulation capacity flow rateCooling coil optional Usable immersion depth 16.6 cm
Catalog No. 50-290-9036