Learn More
Sigma Aldrich Fine Chemicals Biosciences JulaboR CORIOTM CD heating1EA
Catalog No. 502908913
Encompass
The Open Heating Bath Circulators of the new CORIO series feature high-quality bath tanks made of stainless steel and pump connections.Your advantagesModels for internal and external applicationsBright white easy to read displayVery quietEasy pump change-over between internal and external circulationUSB connectionHigh-quality bath tanks made of stainless steelIntegrated drain screw (except B5)Cooling coil and bath cover available as optional accessories
Catalog No. 50-290-8913 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7430161EA
The Open Heating Bath Circulators of the new CORIO series feature high-quality bath tanks made of stainless steel and pump connections.Your advantagesModels for internal and external applicationsBright white easy to read displayVery quietEasy pump change-over between internal and external circulationUSB connectionHigh-quality bath tanks made of stainless steelIntegrated drain screw (except B5)Cooling coil and bath cover available as optional accessories