Learn More
Sigma Aldrich Fine Chemicals Biosciences JulaboR CORIOTM CD refridg1EA
Catalog No. 502908635
Encompass
Refrigerated / Heating Circulators of the new CORIO series distinguish themselves with a great price-to-performance ratio. They are ideal for all standard tasks and routine work in laboratories and industry..nbspModels for internal and external applications.nbspBright white easy to read display.nbspVery quiet.nbspEasy change-over from internal to external circulation and vice versa.nbspExternal pump connections (M16x1).nbspUSB interface.nbspSpace-saving cooling coil design provides more usable space in the bath tank.nbspBath lid and drain tap included.nbspRemovable ventilation grid.nbspRefrigeration unit without side vents.nbspClass III (FL) according to DIN 12876-1
Catalog No. 50-290-8635 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7430571EA
Add to Cart
Add to Cart
Request a Quote
Refrigerated / Heating Circulators of the new CORIO series distinguish themselves with a great price-to-performance ratio. They are ideal for all standard tasks and routine work in laboratories and industry..nbspModels for internal and external applications.nbspBright white easy to read display.nbspVery quiet.nbspEasy change-over from internal to external circulation and vice versa.nbspExternal pump connections (M16x1).nbspUSB interface.nbspSpace-saving cooling coil design provides more usable space in the bath tank.nbspBath lid and drain tap included.nbspRemovable ventilation grid.nbspRefrigeration unit without side vents.nbspClass III (FL) according to DIN 12876-1