Learn More
Sigma Aldrich Fine Chemicals Biosciences JulaboR shaking water bath1EA
Catalog No. 502908772
Encompass
.nbspMicroprocessor technology.nbspSplash-proof mains switch.nbspKeypad for actual setpoint values high and low temperature warning functions.nbspMulti-Display (LED).nbspLarge bath volume.nbspReduced space requirement.nbspRS232 interface.nbspElectronic timerCover needs to be ordered separately.
Catalog No. 50-290-8772 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5573151EA
Add to Cart
Add to Cart
Request a Quote
.nbspMicroprocessor technology.nbspSplash-proof mains switch.nbspKeypad for actual setpoint values high and low temperature warning functions.nbspMulti-Display (LED).nbspLarge bath volume.nbspReduced space requirement.nbspRS232 interface.nbspElectronic timerCover needs to be ordered separately.