Learn More
Sigma Aldrich Fine Chemicals Biosciences JulaboR water baths PURA 21EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7431571EA
Encompass
The PURA water bath from JULABO sets new standards of performance with innovative technologies that leave conventional water baths behind. Users enjoy numerous benefits in their daily work. JULABO engineers prioritized innovative functionality a cutting-edge design and the use of completely novel technologies as early as the conceptualization phase of development. The results are nothing short of brilliant. A PURA water bath will streamline the daily routines in your laboratory with innovative functionality.All PURA water baths have an integrated drain tap. Enameled and glass-hard parts in the bath itself are highly durable and - most importantly - hygienically clean. Polycarbonate sidewalls have better insulating properties than stainless steel and are less conductive but are equal to stainless steel in terms of cleanliness and hygiene. Operating the water bath is very straightforward and the bright white LED display is easy to read even from a distance. USB data communication is
Catalog No. 50-290-8739