Learn More
Thermco Products Inc JUMBO DIGIT THERM/HYGROMETER
Catalog No. NC0886116
Encompass_Preferred
• Temperature Range: -10° to 60°C (-14° to 140°F)• Humidity Range: +10 to 100% RH• Accuracy: ±1.0°C & F and ±2% Mid Range ±4% Elsewhere• Resolution: 0.1 Temperature and 1.0% Relative Humidity• Minimum/Maximum Temperature and Humidity Memory Recalling• Fahrenheit/Celsius Selectable• Flip-out Stand• Low Battery Indicator• Battery Included
Catalog No. NC0886116 Supplier Thermco Products Inc Supplier No. ACCDTH03
• Temperature Range: -10° to 60°C (-14° to 140°F)• Humidity Range: +10 to 100% RH• Accuracy: ±1.0°C & F and ±2% Mid Range ±4% Elsewhere• Resolution: 0.1 Temperature and 1.0% Relative Humidity• Minimum/Maximum Temperature and Humidity Memory Recalling• Fahrenheit/Celsius Selectable• Flip-out Stand• Low Battery Indicator• Battery Included