Learn More
Research Products International Corp Jumbo Display Timer, 5 x 3 3/4 x 1/2 Inches, Battery Included
SDP
Catalog No. 50999168
Encompass
Features the largest display available on a timer: 3 x 2 1/2 Inches. Can be easily read from across a room. Simultaneously displays either one count up channel and a clock, or two count down channels. All channels have a range of 24 hours with 1 second resolution. Also indicates how much time has elapsed after a count down alarm sounds and conveniently recalls the last timer setting for reuse. White case has extra large start/stop button and a loud alarm. Wall mount or fold for bench top use.
Runs for approximately 2 years on one AAA battery (included).
Catalog No. 50-999-168 Supplier Research Products International Corp Supplier No. 810006
Features the largest display available on a timer: 3 x 2 1/2 Inches. Can be easily read from across a room. Simultaneously displays either one count up channel and a clock, or two count down channels. All channels have a range of 24 hours with 1 second resolution. Also indicates how much time has elapsed after a count down alarm sounds and conveniently recalls the last timer setting for reuse. White case has extra large start/stop button and a loud alarm. Wall mount or fold for bench top use.
Runs for approximately 2 years on one AAA battery (included).