Learn More
Research Products International Corp Jumbo Genie Blotter, 25 x 30 cm, with Platinum Anode
SDP
Supplier:  Research Products International Corp 144041
Encompass
Research laboratories worldwide use Genie Blotters for rapid and accurate electrophoretic transfers. The unique design closely positions the plate electrodes to produce high field strengths(in volts/cm) with low voltage power sources. Superb Western Blots can be obtained in as little as 30 minutes at 24 volts. Northern and Southern blots in as little as 60-90 minutes.
GENIE also features an easy to load cassette tray for submerged bubble-free assembly. Slide the loaded tray into the tray holder and stand unit upright. The vertical operation eliminates bubble accumulation, gel slumping and burned blots.
Supplied complete with,platinized titanium anode, power supply (6 and 12 volt, 10 amp output, with timer. 110 volt input) cassette tray, tray holder, electrodes, Scotch-Brite pads, bubble screens and complete instructions. This model with a platinized titanium anode is recommended for blotting high molecular weight proteins or large nucleic acids.
Catalog No. 50-197-6933