Learn More
Grainger MERET MJD Medical Oxygen Cylinder, Aluminum, 623 L, 1 ea
Catalog No. 502783352
Encompass_Preferred
This MJD (Jumbo D or M22) refillable medical oxygen cylinder is constructed from high-strength aluminum 6061-T6 alloy. It is designed for healthcare applications with a 2216 PSI filling pressure and an oxygen capacity of 22 cu ft (623 L). The unit is approved to DOT 3AL and TC 3ALM standards and features a clear powder coat protective finish with a green neck for oxygen identification.
  • Manufactured in the USA
  • DOT 3AL and TC 3ALM approved
  • Features 0.750-16 UNF-2B thread size
  • Equipped with a CGA870 post valve
  • Includes a 5 year hydrostatic test capacity
  • Clear powder coat finish with green neck
  • Overall dimensions of 19.5 in. x 4.38 in. x 4.38 in. with valve
Catalog No. 50-278-3352 Supplier Grainger Supplier No. 19MN02
This MJD (Jumbo D or M22) refillable medical oxygen cylinder is constructed from high-strength aluminum 6061-T6 alloy. It is designed for healthcare applications with a 2216 PSI filling pressure and an oxygen capacity of 22 cu ft (623 L). The unit is approved to DOT 3AL and TC 3ALM standards and features a clear powder coat protective finish with a green neck for oxygen identification.
  • Manufactured in the USA
  • DOT 3AL and TC 3ALM approved
  • Features 0.750-16 UNF-2B thread size
  • Equipped with a CGA870 post valve
  • Includes a 5 year hydrostatic test capacity
  • Clear powder coat finish with green neck
  • Overall dimensions of 19.5 in. x 4.38 in. x 4.38 in. with valve