Learn More
BURKLE INC Jumbo sampler, all layer, Alu, 33.46 in. (85 cm), 29.76 oz. (880 ml) volume
Catalog No. 5000025644
Encompass_Preferred
The Jumbo zone sampler is used for sampling through all layers, especially of coarse-grained bulk materials. The long sampling lances with 50 mm tube diameter and the extra large chamber volume allow the sampling of large quantities of large grain sizes.
This zone sampler ensures representative sampling through all layers or zones of the sample material and is essential for accurate analysis and quality control in various industries.
Because the lances can be used both vertically and horizontally, this sampler offers maximum flexibility when sampling from different types of containers, silos or piles.
Catalog No. 50-000-25644 Supplier BURKLE INC Supplier No. 53191085
The Jumbo zone sampler is used for sampling through all layers, especially of coarse-grained bulk materials. The long sampling lances with 50 mm tube diameter and the extra large chamber volume allow the sampling of large quantities of large grain sizes.
This zone sampler ensures representative sampling through all layers or zones of the sample material and is essential for accurate analysis and quality control in various industries.
Because the lances can be used both vertically and horizontally, this sampler offers maximum flexibility when sampling from different types of containers, silos or piles.