Learn More
Description
- Kids can depict the passing of time in an engaging and calming way. The orange-colored sand is coordinated with the hourglass timer.
- This set includes one 5-minute jumbo sand timer. Watch time pass by with this colorful large sand timer.
- It's sized for little hands, the oversized timers are perfect for use anywhere children need help managing time.
- Place the sand clock sideways to pause the 5 minute timer.
- The top and bottom end cap is squared-off and made from soft rubber that offers increased durability and allows for quiet pausing of the timer.
- The hourglass sand timer is clearly labeled with a specific number on top, which makes it easy to tell apart.
- The blue timer is 1 minute, green is 2 minutes, orange is 5 minutes, and the purple is 10 minutes.
- Help kids develop a stronger sense of time, productivity, and efficiency.
- The perfect timer for teeth cleaning, studying, show and tell, yoga, gym time, and time out.
- Large hourglass sand timer for kids are not precision timing devices, so there is time deviation.
- Students in activity-based programs perform up to 20% higher than groups using traditional or textbook approaches.
- For over 50 years, hand2mind has encouraged hands-on learning and discovery, driving deeper understanding and helping children unlock their full potential. With products designed to enhance learning in the classroom and the world around us, we strive to find new ways to encourage hands-on exploration and discovery.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Class |
| Product Type | Sand Timer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
