Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jumbo Test Tubes with Stand
SCI_ED

Catalog No. s02195
Change view
Click to view available options
Grade:
Pre-K
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02195 Pre-K
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S02195 Supplier Learning Resources, Inc Supplier No. LER2788
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Six test tubes, different colored lids (to track experiments), sturdy rack and Activity Guide

Chunky, durable test tubes put serious science safely within reach for young students.

  • Support inquiry-based investigation and hands-on observation
  • Plastic tubes are safe at temperatures from -12.2° to +93.3°C (10° to 200°F)
  • Tubes feature calibration marks: 25, 50, 75 and 100mL
  • Hand wash

Order Info

Grades PreK+. Size: 1.5D x 6 in. H (4 x 15cm).

Specifications

Grade Level Early Childhood
Grade Pre-K
Dimensions (L x W) 1.5D x 6 in. H (4 x 15 cm)
Includes Six test tubes, different colored lids (to track experiments), sturdy rack and Activity Guide
Material Plastic
Description Jumbo Test Tubes with Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.