Learn More
Sigma Aldrich Fine Chemicals Biosciences JumpStartTM REDTaqR DNA Po
Catalog No. 502907764
Encompass
JumpStart REDTaq DNA Polymerase is Sigmas high performance Taq DNA Polymerase blended with JumpStart Taq antibody and an inert red dye tracer. Extensive testing with a variety of primers and templates indicates that the performance of JumpStart REDTaq DNA Polymerase is equivalent to or better than that of standard Taq polymerase.The hot start Taq antibody prevents non-specific product formation.Assembled PCR reactions can be placed at room temperature up to 2 hours.Red tracer allows visible tracking of enzyme addition and reaction mixing.PCR can be directly loaded onto an agarose gel without addition of loading buffers.The red tracer is a tracking dye that run at 125bp on a 1% agarose gel. Since the red tracer has no effect on the amplification process a sample can be easily re-amplified such as in nested PCR. The presence of the dye also has no effect on automated DNA sequencing ligase mediated ligations exonucleolytic PCR product digestion and transforma
Catalog No. 50-290-7764 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D81872.5KU
JumpStart REDTaq DNA Polymerase is Sigmas high performance Taq DNA Polymerase blended with JumpStart Taq antibody and an inert red dye tracer. Extensive testing with a variety of primers and templates indicates that the performance of JumpStart REDTaq DNA Polymerase is equivalent to or better than that of standard Taq polymerase.The hot start Taq antibody prevents non-specific product formation.Assembled PCR reactions can be placed at room temperature up to 2 hours.Red tracer allows visible tracking of enzyme addition and reaction mixing.PCR can be directly loaded onto an agarose gel without addition of loading buffers.The red tracer is a tracking dye that run at 125bp on a 1% agarose gel. Since the red tracer has no effect on the amplification process a sample can be easily re-amplified such as in nested PCR. The presence of the dye also has no effect on automated DNA sequencing ligase mediated ligations exonucleolytic PCR product digestion and transforma