Learn More
Sigma Aldrich Fine Chemicals Biosciences JumpStartTM Taq DNA Polyme
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D4184250UN
Encompass
JumpStart(TM) Taq DNA Polymerase (JumpStart(TM) Taq) is an optimized blend of Sigmas high-performance Taq DNA Polymerase and JumpStart(TM) Taq antibody.
Catalog No. 50-290-5818