Learn More
Sigma Aldrich Fine Chemicals Biosciences JumpStartTM Taq ReadyMixTM
Catalog No. 502904949
Encompass
JumpStart(TM) Taq ReadyMix(TM) is a ready-to-use 2X master mix that contains JumpStart(TM) Taq DNA polymerase 99% pure dNTPs reaction buffer and JumpStart(TM) Taq antibody. The Taq DNA polymerase is an antibody-inactivated hot-start enzyme. Once the reaction temperature reaches 70C the DNA polymerase-antibody complex dissociates and Taq DNA polymerase activity is restored. This antibody-enzyme complex allows for easy and convenient set-up with less contamination risk than manual hot-start techniques.
Catalog No. 50-290-4949 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P2893400RXN
JumpStart(TM) Taq ReadyMix(TM) is a ready-to-use 2X master mix that contains JumpStart(TM) Taq DNA polymerase 99% pure dNTPs reaction buffer and JumpStart(TM) Taq antibody. The Taq DNA polymerase is an antibody-inactivated hot-start enzyme. Once the reaction temperature reaches 70C the DNA polymerase-antibody complex dissociates and Taq DNA polymerase activity is restored. This antibody-enzyme complex allows for easy and convenient set-up with less contamination risk than manual hot-start techniques.