Learn More
Aviva Systems Biology Corp JUN Antibody - N-terminal region (ARP38133_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38133_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against JUN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human JUN. Alternate names: AP1, AP-1, c-JunApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56529