Learn More
Aviva Systems Biology Corp JUN Antibody - middle region (ARP38134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JUN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human JUN. Alternate names: AP1, c-Jun, AP-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56530