Learn More
Aviva Systems Biology Corp JUNB Antibody - N-terminal region (P100946_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100946_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against JUNB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human JUNB. Alternate names: AP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62794