Learn More
Hach Company Junction box-SD900 4-20 input
Supplier:  Hach Company 8760600
Encompass
4-20mA interface Converts analog signal from flow meter to pulses to make compatible with SD900 sampler
Catalog No. 50-002-07143