Learn More
Aviva Systems Biology Corp Jundm2 Antibody - N-terminal region (ARP37413_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37413_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Jundm2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of mouse Jundm2. Alternate names: Jdp2, Jundp2, TIF, Jundm2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-627