Learn More
Alkali Scientific ASI™ Junior Multi tube Rack, Red, holds 25 Microtubes & 16 x 0.5 or 16 x 0.2 on Reverse, 10/Pk
SDP
Catalog No. 503026924
Encompass_Preferred
This smaller model of the UniRackTM can hold up to 25 polystyrene or polypropylene 10 and 12 mm tubes, such as 10 x 75 mm or 12 x 75 mm sizes. This rack will also accommodate all types of screw cap microtubes from 0.5 to 2 ml as well as 1 to 5 ml cryogenic vials. Flip the UniRackTM over and you can store up to 16 PCR or microcentrifuge tubes sized from 0.2 to 0.5 ml. 
Catalog No. 50-302-6924 Supplier Alkali Scientific Supplier No. MTR01R
This smaller model of the UniRackTM can hold up to 25 polystyrene or polypropylene 10 and 12 mm tubes, such as 10 x 75 mm or 12 x 75 mm sizes. This rack will also accommodate all types of screw cap microtubes from 0.5 to 2 ml as well as 1 to 5 ml cryogenic vials. Flip the UniRackTM over and you can store up to 16 PCR or microcentrifuge tubes sized from 0.2 to 0.5 ml. 