Learn More
MSC Honeywell Junk Yard Dog Cut-Resistant Gloves, Kevlar, Leather Palm, 1 Pair
Catalog No. 5000508902
Encompass
These cut-resistant gloves feature a 100% Kevlar® shell with a premium leather palm for reliable hand protection. Designed for demanding environments, the gloves offer a combination of flame, heat, and abrasion resistance while maintaining an improved grip in wet or oily conditions.
  • Seamless knit construction for comfort and reduced chafing
  • Full premium leather palm and thumb crotch for durability and grip
  • Heat resistance up to 482°C (900°F) without fiber degradation
  • ANSI/ISEA cut level A3 rating
  • ASTM 3389-05 abrasion resistance level 6
  • Launderable without shrinkage
Catalog No. 50-005-08902 Supplier MSC Supplier No. 05831466
These cut-resistant gloves feature a 100% Kevlar® shell with a premium leather palm for reliable hand protection. Designed for demanding environments, the gloves offer a combination of flame, heat, and abrasion resistance while maintaining an improved grip in wet or oily conditions.
  • Seamless knit construction for comfort and reduced chafing
  • Full premium leather palm and thumb crotch for durability and grip
  • Heat resistance up to 482°C (900°F) without fiber degradation
  • ANSI/ISEA cut level A3 rating
  • ASTM 3389-05 abrasion resistance level 6
  • Launderable without shrinkage