Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3728MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CTNG/1664. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: A431, T47D or MCF-7 cells. Skin. Cellular Staining: Cell Surface and Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length gamma catenin protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 80-87kDa.Chromosome: 17q21. Gene Symbol: JUP. Gene ID: 3728. Ensemble: ENSG00000173801. Uniprot: NP_001339702.
Catalog No. 50-188-4848