Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Jupiter 5 µm C4 300 Å

Catalog No. 502395805 Shop All Phenomenex Products
Encompass

Jupiter 5 µm C4 300 Å LC Column 250 x 30 mm

Catalog No. 50-239-5805 Supplier Phenomenex Inc Supplier No. 00G4167U0
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Jupiter 5 µm C4 300 Å LC Column 250 x 30 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.