Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Jupiter 5 µm C4 300 Å

Catalog No. 502394458 Shop All Phenomenex Products
Encompass

Jupiter 5 µm C4 300 Å LC Column 150 x 30 mm, AXIA™ Packed

Catalog No. 50-239-4458 Supplier Phenomenex Inc Supplier No. 00F4167U0AX
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Jupiter 5 µm C4 300 Å LC Column 150 x 30 mm, AXIA™ Packed

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.