Learn More
Eurofins Discoverx PathHunter Jurkat CD47-Presenting Cell Line, 2 vials. (93-1149C19)
Catalog No. 502002257
Encompass
The PathHunter CD47 Ligand cell line is a stable, engineered cell line designed for use with SIRPα Checkpoint Signaling Cell Lines. When co-cultured, the CD47 expressed in this cell line will induce activity of SIRPα receptors, which may be assayed through SH2 domain protein recruitment. This product is a companion product designed to work with Immunoglobulin Superfamily Checkpoint Assays. The cell line included overexpresses CD47 cell surface ligand. When co-cultured with the SIRPa Checkpoint Signaling Cell Line, the CD47 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.
Catalog No. 50-200-2257 Supplier Eurofins Discoverx Supplier No. 931149C19
Add to Cart
Add to Cart
The PathHunter CD47 Ligand cell line is a stable, engineered cell line designed for use with SIRPα Checkpoint Signaling Cell Lines. When co-cultured, the CD47 expressed in this cell line will induce activity of SIRPα receptors, which may be assayed through SH2 domain protein recruitment. This product is a companion product designed to work with Immunoglobulin Superfamily Checkpoint Assays. The cell line included overexpresses CD47 cell surface ligand. When co-cultured with the SIRPa Checkpoint Signaling Cell Line, the CD47 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.