Learn More
ATCC Jurkat, Clone E6-1, Acute T Cell Leukemia, Frozen
Supplier:  ATCC TIB152/DEL
Encompass
Clone E6-1 cells produce large amounts of IL-2 after stimulation with phorbol esters and either lectins or monoclonal antibodies against the T3 antigen (both types of stimulants are needed to induce IL-2 production. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-852FP