Learn More
ATCC Jurkat, Clone E6-1; Acute T Cell Leukemia; Human (Homo sapiens)
Catalog No. 502382564
Encompass
Jurkat, Clone E6-1 is a clone of the Jurkat-FHCRC cell line, a derivative of the Jurkat cell line, which was established from the peripheral blood of a 14-year-old, male, acute T-cell leukemia patient. This cell line can be used in immune system disorder research and immunology and immuno-oncology research.
Catalog No. 50-238-2564 Supplier ATCC Supplier No. TIB152
Jurkat, Clone E6-1 is a clone of the Jurkat-FHCRC cell line, a derivative of the Jurkat cell line, which was established from the peripheral blood of a 14-year-old, male, acute T-cell leukemia patient. This cell line can be used in immune system disorder research and immunology and immuno-oncology research.