Learn More
Invivogen ADCP Reporter Cells - Human T Lymphocytes, 3-7 x 10e6 cells
SDP
Catalog No. NC1982137
Encompass
InvivoGen offers a series of Jurkat-Lucia NFAT cell lines, specifically designed to assess the potency of specific immunoglobulin for ADCC (antibody-dependent cellular cytotoxicity) and ADCP (antibody-dependent cell-mediated phagocytosis).
Catalog No. NC1982137 Supplier Invivogen Supplier No. JKTLNFATCD32
InvivoGen offers a series of Jurkat-Lucia NFAT cell lines, specifically designed to assess the potency of specific immunoglobulin for ADCC (antibody-dependent cellular cytotoxicity) and ADCP (antibody-dependent cell-mediated phagocytosis).