Learn More
Eurofins Discoverx PathHunter Jurkat NFAT Pathway Reporter Cell Line, 2 vials. (93-1141C19)
Catalog No. 502002223
Encompass
The PathHunter Jurkat NFAT Pathway Reporter Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting the NFAT pathway. This cell line enables assessment of NFAT pathway stimulation based on changes in the expression of a NFAT response element-driven reporter protein. This product uses EFC technology. The included cell line expresses ProLabel-tagged reporter protein under the control of a NFAT pathway-inducible transcriptional response element. Pathway activation induces expression of the ProLabel-tagged protein, allowing detection of EFC signal upon addition of EA-containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-2223 Supplier Eurofins Discoverx Supplier No. 931141C19
Add to Cart
Add to Cart
Request a Quote
The PathHunter Jurkat NFAT Pathway Reporter Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting the NFAT pathway. This cell line enables assessment of NFAT pathway stimulation based on changes in the expression of a NFAT response element-driven reporter protein. This product uses EFC technology. The included cell line expresses ProLabel-tagged reporter protein under the control of a NFAT pathway-inducible transcriptional response element. Pathway activation induces expression of the ProLabel-tagged protein, allowing detection of EFC signal upon addition of EA-containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.