Learn More
Eurofins Discoverx PathHunter Jurkat PD-1 Signaling Assay (SHP-2), 2 vials. (93-1106C19)
Catalog No. 502002118
Encompass
The PathHunter PD-1 Checkpoint Signaling cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting signaling checkpoints. This cell line enables assessment of ligand (e.g. PD-L1, PD-L2) based activation of PD-1 receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged PD-1 target protein and EA-tagged SH2 domain protein. Upon activation of the receptor by its ligand or an agonist antibody, recruitment of SH2 domain protein to the phosphorylated receptor tail leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.
Catalog No. 50-200-2118 Supplier Eurofins Discoverx Supplier No. 931106C19
Add to Cart
Add to Cart
Request a Quote
The PathHunter PD-1 Checkpoint Signaling cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting signaling checkpoints. This cell line enables assessment of ligand (e.g. PD-L1, PD-L2) based activation of PD-1 receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged PD-1 target protein and EA-tagged SH2 domain protein. Upon activation of the receptor by its ligand or an agonist antibody, recruitment of SH2 domain protein to the phosphorylated receptor tail leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.