Learn More
Braintree Scientific Just for Mouse Stereotaxic Ultra Precise Digital
SDP
Catalog No. 501954087
Encompass
The Just for Mouse™ Stereotaxic Instrument was specifically designed for use with knock-out and transgenic mice. It is our smallest and most economic stereotaxic instrument. Available as Dual, Ultra-Precise, Digital or Motorized. 25cm X 25cm Footprint Gas Anesthesia compatible Ear and tooth bar post height adjustment to accommodate animals between 10-75g Delrin ear bars, with 3 types of head holders Triple lead screw in manipulator arm for fast positioning. Precision alignment when using the Just For Mouse ensures accurate placement of electrodes, micropipettes, cannula, and other devices. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4087 Supplier Braintree Scientific Supplier No. 51730UD
Add to Cart
Add to Cart
Request a Quote
The Just for Mouse™ Stereotaxic Instrument was specifically designed for use with knock-out and transgenic mice. It is our smallest and most economic stereotaxic instrument. Available as Dual, Ultra-Precise, Digital or Motorized. 25cm X 25cm Footprint Gas Anesthesia compatible Ear and tooth bar post height adjustment to accommodate animals between 10-75g Delrin ear bars, with 3 types of head holders Triple lead screw in manipulator arm for fast positioning. Precision alignment when using the Just For Mouse ensures accurate placement of electrodes, micropipettes, cannula, and other devices. **Sold for research purposes only and not for human use**