Learn More
MSC Justrite 1 gal. Steel Plunger Can, Perforated Screen, 1 ea
Catalog No. 5000793209
Encompass
This reliable 1-gal. (4 L) plunger dispensing can, made of 24-gauge steel, is designed to safely and easily moisten cleaning cloths with liquid solvents. The can measures 10.5 in. in height and 7.25 in. in outer diameter, with an accompanying steel dasher plate of 5.0 in.
  • Chemically resistant 'Smart Pump' mechanism for efficient dispensing
  • Perforated pan screen serves as a flame arrester for safety
  • Molded edge protectors on top rim for smooth moistening
  • Raised bottom ensures stability of the can
  • Capacity is 1 gal. (4 L)
  • FM Approved and TUV Certified
  • Leaves less liquid at bottom, reducing refilling frequency
  • Increases saturation with fewer pump strokes
  • Provides a safer method for wetting cloths in various industries
  • Constructed from 24-gauge steel for durability
Catalog No. 50-007-93209 Supplier MSC Supplier No. 89848212
This reliable 1-gal. (4 L) plunger dispensing can, made of 24-gauge steel, is designed to safely and easily moisten cleaning cloths with liquid solvents. The can measures 10.5 in. in height and 7.25 in. in outer diameter, with an accompanying steel dasher plate of 5.0 in.
  • Chemically resistant 'Smart Pump' mechanism for efficient dispensing
  • Perforated pan screen serves as a flame arrester for safety
  • Molded edge protectors on top rim for smooth moistening
  • Raised bottom ensures stability of the can
  • Capacity is 1 gal. (4 L)
  • FM Approved and TUV Certified
  • Leaves less liquid at bottom, reducing refilling frequency
  • Increases saturation with fewer pump strokes
  • Provides a safer method for wetting cloths in various industries
  • Constructed from 24-gauge steel for durability