Learn More
Justrite™ EN Safety Cabinets for Flammables
GSA_VAGREENER_CHOICE
Catalog No. p-6731134 Shop All Justrite Safety Group Products
Change view
Click to view available options
Load Bearing Capacity:
30 gal.
45 gal.
No. of Doors:
1
2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity No. of Doors
19-414-050 30 gal. 1
19-414-051 45 gal. 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-050 Supplier Justrite™ Supplier No. 22601
Only null left
Add to Cart
Add to Cart
Request a Quote
Provides 30 minutes of fire resistance with safe storage and segregation of flammables
  • Attractive two-toned yellow cabinet
  • Constructed with body of melamine resin and flame retardant special plates; Steel door front covered by textured, durable powder-coat finish
  • Provides 30 minutes of content protection and extended evacuation time Hybrid style door open or close manually and include damper that adjusts force needed to move doors
  • Under fire conditions, intumescent strips expand and hermetically seal all gaps to reduce heat entering cabinet and built-in fusible link system releases to allow automatic closure of doors at 122°F (50°C)
  • Powder-coat finish steel shelves adjust on 32mm (1.25 in.) increments with load capacity of 75kg (165 lb)
  • Shelves with high ledge prevent small containers from sliding off
  • Removable bottom sump with detachable perforated insert features three-sided lip seal to capture leaks or spills from shelves at bottom of sump, keeping containers in sump clean and dry
Certifications
Tested and GS certified by MPA Dresden
Compliance
EN 14470-1 / EN 14727 compliant and meet NFPA 30, NFPA 1, and OSHA standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
No. of Doors 1
Load Bearing Capacity 30 gal.
Description 30 minutes fire resistance; Melamine resin, flame-resistant special plates
Material Melamine Resin