Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Base Cover for 30-Minute EN Cabinet

Catalog No. 19414059 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
For 30 gal. EN cabinet
For 45 gal. EN cabinet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-414-059 For 45 gal. EN cabinet
19-414-058 For 30 gal. EN cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-414-059 Supplier Justrite™ Supplier No. 22635
  • $277.00
    $187.65 / Each of 1

    Save $89.35

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keeps debris out from underneath cabinets by adding an optional, removeable base cover

  • Made from melamine resin laminated chipboard
  • Conceals integrated transport base on cabinet
  • Prevents dust and debris from collecting underneath cabinet
  • Creates seamless flush-to-floor attractive presentation
  • Yellow

Specifications

For Use With (Equipment) For 45 gal. EN cabinet
Description Prevents collection of dust, debris; Melamine resin laminated chipboard
Color Yellow
Material Melamine Resin, Chipboard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.