Learn More
MSC Justrite Bench Can, 2 gal, Coated Steel, Red, 1 ea
Catalog No. 5000782878
Encompass
This bench can allows users to place work on a spring-loaded, perforated dasher plate, press down to agitate, and then release to return the dasher to the drain position.
  • Plated steel dasher acts as an effective flame arrester to reduce the risk of fire flashback
  • Hinged cover minimizes evaporation losses and a protected rim reduces the possibility of hand injury
  • Constructed from sturdy 24-gauge steel with a tough powder-coat finish
Catalog No. 50-007-82878 Supplier MSC Supplier No. 89848261
This bench can allows users to place work on a spring-loaded, perforated dasher plate, press down to agitate, and then release to return the dasher to the drain position.
  • Plated steel dasher acts as an effective flame arrester to reduce the risk of fire flashback
  • Hinged cover minimizes evaporation losses and a protected rim reduces the possibility of hand injury
  • Constructed from sturdy 24-gauge steel with a tough powder-coat finish