Learn More
Justrite™ Blue Steel Safety Cabinet
SCI_ED
Catalog No. S28752 Shop All Justrite Safety Group Products
Change view
Click to view available options
Dimensions (D x W x H):
34 x 34 x 65 in.
Catalog No. Dimensions (D x W x H)
S28752 34 x 34 x 65 in.
1 options
Catalog No. S28752 Supplier Justrite™ Supplier No. 896002/EMD
Safely storage acids and other corrosive or hazardous materials. The Justrite™ features double-wall construction, dual vents, and ground wire connections. Epoxy baked-on powder coat finish and paint resists acids, bases, and solvents.
  • Features double-wall construction, dual vents, grounding wire connections
  • Adjustable shelves, leakproof sills, three point self-latching doors and leveling feet
  • Acid cabinets contains polyethylene trays attached to galvanized steel shelves to resist aggressive chemicals
  • Polyethylene liner at bottom sump
  • Epoxy baked-on powder coat finish
  • Paint resists acids, bases and solvents
Specifications
Dimensions (D x W x H) 34 x 34 x 65 in.