Learn More
Justrite™ Brass Faucet
Catalog No. 19018573 Shop All Justrite Safety Group Products
Change view
Click to view available options
Material:
Brass
Catalog No. Material
19-018573 Brass
1 options
Catalog No. 19-018573 Supplier Justrite™ Supplier No. 08540
  • $116.00
    $109.65 / Each of 1
    Save $6.35
    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Self-closing safety faucets prevents flashback of fire into drum
  • Features polytetrafluoroethylene (PTFE) seal
  • Adjustable models screw tightly into drum
  • Faucet extensions provide extra dispensing accuracy and prevents spills
Certifications
FM Global tested and approved
Specifications
Certifications/Compliance FM
Product Type Brass Control Flow Lab Safety Faucet
Closure Type Self Close
Material Brass
Inlet Connection NPTF (English) 0.75 in.
Inlet Connection NPTF (Metric) 1.90 cm