Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Chemcor™ Lined Slimline Safety Cabinets for Hazardous Materials
GREENER_CHOICE

Catalog No. 19414817 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-414-817 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-414-817 Supplier Justrite™ Supplier No. 8622282
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Specifications

Product Type Combi Safety Storage Cabinet
Dimensions (D x W x H) Interior 14.6 x 20 x 60.5 in.
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Door Style Self-close
Load Bearing Capacity 22 gal. (83 L)
No. of Shelves 3
Surface Coating Powder-coated
Shelf Type Adjustable
Weight (Metric) 87.1 kg
Weight (English) 192 lb.
Certifications/Compliance International Fire Code, NFPA Code 30, NFPA 1 Fire-Code, NFPA 400
Material Steel (CR)
Color Blue
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.