Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ ChemCor™ Piggyback Hazardous Material Safety Cabinet
GREENER_CHOICE

Catalog No. 19414819 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Royal Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-414-819 Royal Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-414-819 Supplier Justrite™ Supplier No. 8613282
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Double-wall, 1mm (18 ga) welded steel construction, 51mm (2 in.) liquid-tight containment sump, dual vents, adjustable shelves, three-point stainless steel bullet self-latching doors with U-Loc™ padlockable handle and Haz-Alert™ reflective warning labels, polyethylene work tray, Hazardous - Keep Fire Away label, pack of ten application-specific labels to choose from and apply to cabinet in order to indicate its contents

Expands storage and safely segregates hazardous materials

  • Provides complete protection from harsh acids and corrosive chemicals
  • Designed to mount on top of our standard 30- and 45-gal. (114- and 170-L) safety cabinets
  • Cabinets can also be used independently as wall-mount unit or on countertop
  • ChemCor thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals
  • Seam-free surface eliminates need for rivets or fasteners which risk corrosion from seep-through chemicals
  • Non-porous surface resists staining and cleans up easily
  • All epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance
  • Self-close door shuts and latches automatically when fusible link melts at 74°C (165°F) under fire conditions concealed self-closing mechanism offers obstruction-free access to contents

Warranty and Services

Ten-year warranty

Specifications

Load Bearing Capacity 45 L (24 gal.)
Product Type Piggyback hazardous material safety cabinet
Door Style Self-close
Width (Metric) Exterior 109.2 cm
Depth (Metric) Exterior 45.7 cm
Description Cold-rolled Steel (18 ga)
Dimensions (D x W x H) Interior 37 x 100.3 x 33 cm (14.5 x 39.5 x 13 in.)
Height (Metric) Exterior 45.7 cm
No. of Doors 2
Width (English) Exterior 43 in.
Depth (English) Exterior 18 in.
Color Royal Blue
Certifications/Compliance FM approved, NFPA 1, NFPA 400 Hazardous Material Code, IFC compliant
Height (English) Exterior 18 in.
Material Steel (CR)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.