Description
This fitting disconnect assembly is designed for use with Justrite HPLC waste disposal safety cans.
- Material: Stainless steel with EPDM seal
- Tubing ID: 1/4 in. (6 mm)
- Design: Quick-disconnect assembly
- Installation: Pre-installed
Specifications
Specifications
| Includes | EPDM seal |
| Material | Stainless Steel |
| Product Type | Fitting |