Learn More
MSC Justrite Drum Faucet, Brass, 3/4 in. NPT, Viscous, 1 ea
Catalog No. 5000675396
Encompass
These faucets are designed for leakproof dispensing of flammable liquids from drums. They feature an internal flame arrester to prevent flashback of fire into the drum, enhancing safety. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Ideal for viscous liquids
  • Brass construction
  • Manual close for dispensing
  • Safe and easy dispensing from horizontal drums
  • FM approved
Catalog No. 50-006-75396 Supplier MSC Supplier No. 89847701
These faucets are designed for leakproof dispensing of flammable liquids from drums. They feature an internal flame arrester to prevent flashback of fire into the drum, enhancing safety. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Ideal for viscous liquids
  • Brass construction
  • Manual close for dispensing
  • Safe and easy dispensing from horizontal drums
  • FM approved