Learn More
Justrite™ Drum Faucets
Catalog No. 19812450 Shop All Justrite Safety Group Products
Change view
Click to view available options
Closure Type:
Manual
Catalog No. Closure Type
19-812-450 Manual
1 options
Catalog No. 19-812-450 Supplier Justrite™ Supplier No. 08955
Add to Cart
Add to Cart
For viscous liquids
  • Gasketed to prevent leakage
  • Self-closing
  • Made of brass
  • Internal flame arrester
  • 3/4 in. NPT bung
Specifications
Product Type Drum Faucet
Closure Type Manual
For Use With (Equipment) For Viscous Liquids