Learn More
Justrite™ Drum Faucets, Extender, and Valve
Catalog No. 19018575 Shop All Justrite Safety Group Products
Change view
Click to view available options
Product Type:
Adjustable Drum Faucet
Drum Gate Valve
Flexible Hose Extension
Rigid Drum Faucet
Material:
Brass
Stainless Steel
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
19-018575 Flexible Hose Extension Stainless Steel
17-185C Flexible Hose Extension Brass
17-185D Drum Gate Valve Brass
17-986-53 Adjustable Drum Faucet Brass
17-986-54 Rigid Drum Faucet Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-018575 Supplier Justrite™ Supplier No. 08587
Add to Cart
Add to Cart
Self-closing safety drum faucets
  • Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
  • Brass faucets screw into drum; adjustable model adjusts to spout-down position
  • Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
  • Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifications
Certifications/Compliance FM
Product Type Flexible Hose Extension
For Use With (Application) Provide Extra Accuracy and Prevent Spills
For Use With (Equipment) 08916 Faucet
Material Stainless Steel
Height (English) 6 in.
Height (Metric) 15.2 cm