Learn More
Justrite™ Drum Lifter
Catalog No. 17188 Shop All Justrite Safety Group Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
17-188 Steel
1 options
Catalog No. 17-188 Supplier Justrite™ Supplier No. 08801
  • $530.00
    $318.65 / Each of 1
    Save $211.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Can be used with a forklift or overhead pulley
  • Sturdy heavy-gauge steel drum lifter attaches securely to any drum rim
  • Holds up to 1000 lb. (454kg)
Specifications
Product Type Tong Style Drum Lifter
For Use With (Application) Allowing a 55 gal. Steel drum to be lifted and moved with a forklift truck
For Use With (Equipment) Forklift or Overhead Pulley
Material Steel