Learn More
Justrite™ Drum Siphon Adapter
Catalog No. 19041647 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30 and 55 gal. Draining Drums
Catalog No. For Use With (Equipment)
19-041-647 30 and 55 gal. Draining Drums
1 options
Catalog No. 19-041-647 Supplier Justrite™ Supplier No. 08311
  • $328.00
    $196.65 / Each of 1
    Save $131.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For draining 30 and 55 gallon drums
  • One end of the adapter accepts a standard 1.9cm (0.75 in.) drum faucet
  • Opposite end screws into a 1.9cm (0.75 in.) drum bung opening
  • Adjustable collar
Specifications
Product Type Drum Siphon Adapter
For Use With (Application) Empties residual solvents from a typical two to three gallons down to the last few ounces, eliminating the need for dangerous drum tipping or rolling
For Use With (Equipment) 30 and 55 gal. Draining Drums
Material Brass
Includes Adjustable Collar