Learn More
Justrite™ EL Fitting for Horizontal and Vertical Drum Vents
Catalog No. 19018555 Shop All Justrite Safety Group Products
Change view
Click to view available options
Material:
Non-metallic Cast Iron
Catalog No. Material
19-018555 Non-metallic Cast Iron
1 options
Catalog No. 19-018555 Supplier Justrite™ Supplier No. 08015
Add to Cart
Add to Cart
Cast iron EL fitting for use with Justrite Horizontal and Vertical Drum Vents
  • Used for mounting 08101 or 08005 vents in 2cm (0.75 in.) end drum opening
  • Cast iron
Order Info
For drum vents, see cat. no. 17186A.
Certifications
FM approved.
Specifications
Certifications/Compliance FM
Product Type El Fitting
For Use With (Application) Controlled release of flammable vapors, prevent explosions caused by pressure buildup
For Use With (Equipment) Mounting drum vents 17-186A and 17-186B in 0.75 in. end drum opening
Material Non-metallic Cast Iron