Learn More
MSC Justrite Fire Resistant Steel Drum, 4.50 gal., UL, 1 ea
Catalog No. 5000765955
Encompass
This fire resistant steel drum is designed to extinguish internal fires by cutting off the air supply. It is intended for collecting waste paper and other solvent-free combustibles, providing continuous protection against trash fires in various environments.
  • Designed to extinguish internal fires by cutting off air supply.
  • Provides continuous protection against trash fires.
  • Suitable for collecting waste paper and other solvent-free combustibles.
  • Ideal for use in plants, hospitals, schools, offices, and stores.
  • Available with durable steel or brushed aluminum drum heads.
Catalog No. 50-007-65955 Supplier MSC Supplier No. 89848428
This fire resistant steel drum is designed to extinguish internal fires by cutting off the air supply. It is intended for collecting waste paper and other solvent-free combustibles, providing continuous protection against trash fires in various environments.
  • Designed to extinguish internal fires by cutting off air supply.
  • Provides continuous protection against trash fires.
  • Suitable for collecting waste paper and other solvent-free combustibles.
  • Ideal for use in plants, hospitals, schools, offices, and stores.
  • Available with durable steel or brushed aluminum drum heads.