Learn More
MSC Justrite Flammable Cabinet, 12 gal., Manual Close, 1 ea
Catalog No. 5000766612
Encompass
The Sure-Grip® EX Compac Safety Cabinet, with manual close door, provides safe, code-compliant storage for flammable liquids on countertops. Portable, fire-resistant, leak-tested, it meets OSHA, NFPA 30/1 standards, and is FM-approved.
  • Fire-resistant, leak-tested design
  • Sturdy 18-gauge steel double-wall, 1.5 in (38 mm) insulating air space
  • Fully welded construction for durability
  • Manual-close, self-latching door opens 180 degrees
  • U-Loc™ handle with cylinder lock or padlock option
  • Haz-Alert™ reflective warning labels
  • Galvanized steel SpillSlope® shelf, 2-in (51-mm) leak-tested sump
  • Adjustable shelves on 3-in (76-mm) centers, ANSI MH28.1 compliant
  • Dual vents with flame arresters and grounding connector
  • Durable lead-free epoxy/polyester powder-coat finish
  • Includes a 10-year warranty
Catalog No. 50-007-66612 Supplier MSC Supplier No. 71853014
The Sure-Grip® EX Compac Safety Cabinet, with manual close door, provides safe, code-compliant storage for flammable liquids on countertops. Portable, fire-resistant, leak-tested, it meets OSHA, NFPA 30/1 standards, and is FM-approved.
  • Fire-resistant, leak-tested design
  • Sturdy 18-gauge steel double-wall, 1.5 in (38 mm) insulating air space
  • Fully welded construction for durability
  • Manual-close, self-latching door opens 180 degrees
  • U-Loc™ handle with cylinder lock or padlock option
  • Haz-Alert™ reflective warning labels
  • Galvanized steel SpillSlope® shelf, 2-in (51-mm) leak-tested sump
  • Adjustable shelves on 3-in (76-mm) centers, ANSI MH28.1 compliant
  • Dual vents with flame arresters and grounding connector
  • Durable lead-free epoxy/polyester powder-coat finish
  • Includes a 10-year warranty