Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ FM-Approved Biohazard Waste Container
SCI_ED

Catalog No. S04777MF Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
10 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
S04777MF 10 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S04777MF Supplier Justrite™ Supplier No. 05930/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For safe, temporary collection of biological waste.

Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.

  • Leakproof, sturdy steel construction with foot-operated, self-closing lid
  • Eliminate any reason to reach into waste area
  • Hold biohazard bag (sold separately) securely around circumference of can top
  • Large biohazard label clearly identifies the contents
  • Capacity: 10 gal. (38L)

Not to be used for disposal of sharp items.

Certifications

FM approved

Compliance

Complies with OSHA 29CFR1910.1030.

Specifications

Certifications/Compliance FM, OSHA 29 CFR 1910.1030
Diameter (Metric) Outer 35.5 cm
Diameter (English) Outer 14 in.
Capacity (English) 10 gal.
Color White
Height (Metric) 45.7 cm
Includes Foot-operated cover
For Use With (Application) For safe, temporary collection of biological waste
Height (English) 18 in.
Material Metal
Product Type Biohazard Waste Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.